Product Details

SNP ID
rs201478589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120322209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGATATTCAACTCTGACAATAC[C/T]TCTTGGTGTCCAGGTTCTTGTGTGC
Phenotype
MIM: 172410 MIM: 604482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G1B PubMed Links

Gene Details

Gene
PLA2G1B
Gene Name
phospholipase A2 group IB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000928.2 467 Missense Mutation AAG,AGG K144R NP_000919.1
Gene
SIRT4
Gene Name
sirtuin 4
There are no transcripts associated with this gene.

View Full Product Details