Product Details
- SNP ID
-
rs202210528
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:91399126 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGGGTCCGCCCGCAGCGAGGGCG[A/T]GCAGACCTGCGAAGAAGTTCGGAAA
- Phenotype
-
MIM: 602446
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPC5
PubMed Links
Gene Details
- Gene
- GPC5
- Gene Name
- glypican 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004466.5 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
NP_004457.1 |
XM_011521054.2 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_011519356.1 |
XM_011521055.2 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_011519357.1 |
XM_011521056.2 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_011519358.1 |
XM_011521057.2 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_011519359.1 |
XM_011521058.2 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_011519360.1 |
XM_011521059.2 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_011519361.1 |
XM_011521060.2 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_011519362.1 |
XM_017020435.1 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_016875924.1 |
XM_017020436.1 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_016875925.1 |
XM_017020437.1 |
440 |
Missense Mutation |
GAG,GTG |
E27V |
XP_016875926.1 |
View Full Product Details