Product Details

SNP ID
rs201705606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94440055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTACTAAGAGAGTTGTGGGCCAA[A/C]CTGGAGTTTCTTCAACTGAAACTAA
Phenotype
MIM: 191275
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCT PubMed Links

Gene Details

Gene
DCT
Gene Name
dopachrome tautomerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129889.2 1830 Missense Mutation NP_001123361.1
NM_001322182.1 1830 Missense Mutation NP_001309111.1
NM_001322183.1 1830 Missense Mutation NP_001309112.1
NM_001322184.1 1830 Missense Mutation NP_001309113.1
NM_001322185.1 1830 Missense Mutation NP_001309114.1
NM_001322186.1 1830 Missense Mutation NP_001309115.1
NM_001922.4 1830 Missense Mutation NP_001913.2
XM_011521049.1 1830 Missense Mutation XP_011519351.1
XM_017020401.1 1830 Missense Mutation XP_016875890.1

View Full Product Details