Product Details

SNP ID
rs199943435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27273274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATTCAGCTAGCCAATGAGCTGG[A/G]CAGCCTGTTCCTTGAAATTTCCACT
Phenotype
MIM: 612403
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RASL11A PubMed Links

Gene Details

Gene
RASL11A
Gene Name
RAS like family 11 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206827.1 1127 Missense Mutation GAC,GGC D170G NP_996563.1

View Full Product Details