Product Details
- SNP ID
-
rs200369995
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:108208751 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAACCGAAGCTCCAAGGCTTTAA[C/T]AGCTAACCTTGTCCCCTCATTTTTG
- Phenotype
-
MIM: 601837
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ABHD13
PubMed Links
Gene Details
- Gene
- ABHD13
- Gene Name
- abhydrolase domain containing 13
There are no transcripts associated with this gene.
- Gene
- LIG4
- Gene Name
- DNA ligase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098268.1 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
NP_001091738.1 |
NM_002312.3 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
NP_002303.2 |
NM_206937.1 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
NP_996820.1 |
XM_005254056.1 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_005254113.1 |
XM_005254057.4 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_005254114.1 |
XM_005254058.3 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_005254115.1 |
XM_006719951.3 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_006720014.1 |
XM_006719952.1 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_006720015.1 |
XM_011521091.2 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_011519393.1 |
XM_011521092.2 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_011519394.1 |
XM_017020563.1 |
3103 |
Missense Mutation |
ATT,GTT |
I852V |
XP_016876052.1 |
XM_017020564.1 |
3103 |
Missense Mutation |
ATT,GTT |
I852V |
XP_016876053.1 |
XM_017020565.1 |
3103 |
Missense Mutation |
ATT,GTT |
I852V |
XP_016876054.1 |
XM_017020566.1 |
3103 |
Missense Mutation |
ATT,GTT |
I852V |
XP_016876055.1 |
XM_017020567.1 |
3103 |
Missense Mutation |
ATT,GTT |
I852V |
XP_016876056.1 |
XM_017020568.1 |
3103 |
Missense Mutation |
ATT,GTT |
I844V |
XP_016876057.1 |
XM_017020569.1 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_016876058.1 |
XM_017020570.1 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_016876059.1 |
XM_017020571.1 |
3103 |
Missense Mutation |
ATT,GTT |
I840V |
XP_016876060.1 |
XM_017020572.1 |
3103 |
Missense Mutation |
ATT,GTT |
I773V |
XP_016876061.1 |
XM_017020573.1 |
3103 |
Missense Mutation |
ATT,GTT |
I773V |
XP_016876062.1 |
View Full Product Details