Product Details

SNP ID
rs199601303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:40754030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCTTAACAAACCTGTTTCAGGT[G/T]CTTCTTTAGTAACTGCCATCATGTC
Phenotype
MIM: 611992
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPS31 PubMed Links

Gene Details

Gene
MRPS31
Gene Name
mitochondrial ribosomal protein S31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005830.3 878 Missense Mutation GAA,GCA E268A NP_005821.2

View Full Product Details