Product Details
- SNP ID
-
rs199923188
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:89162581 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCGGTGCGTCTCTTTTTGAGGG[A/G]CAGTGTGTCGCTGGGGACCTTCCTG
- Phenotype
-
MIM: 602628
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXN3
PubMed Links
Gene Details
- Gene
- FOXN3
- Gene Name
- forkhead box N3
View Full Product Details