Product Details

SNP ID
rs199676704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100538744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGTAGGTGGTGGCGGGTGGCTTG[C/T]GGTCGAAGAGCTCGTCGCGGCTGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BEGAIN PubMed Links

Gene Details

Gene
BEGAIN
Gene Name
brain enriched guanylate kinase associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159531.1 1295 Missense Mutation CAC,CGC H336R NP_001153003.1
NM_020836.3 1295 Missense Mutation CAC,CGC H336R NP_065887.1
XM_005267919.3 1295 Missense Mutation CAC,CGC H349R XP_005267976.1
XM_005267920.4 1295 Missense Mutation CAC,CGC H348R XP_005267977.1
XM_005267921.3 1295 Missense Mutation CAC,CGC H336R XP_005267978.1
XM_011537031.1 1295 Missense Mutation CAC,CGC H355R XP_011535333.1
XM_017021521.1 1295 Missense Mutation CAC,CGC H336R XP_016877010.1
XM_017021522.1 1295 Missense Mutation CAC,CGC H327R XP_016877011.1
Gene
WDR25
Gene Name
WD repeat domain 25
There are no transcripts associated with this gene.

View Full Product Details