Product Details
- SNP ID
-
rs200427110
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:36477005 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATGAACTCCTGCTTTGTGACAGC[A/G]TCGTGGGATGCAGCAGGGCTGTATG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SFTA3
PubMed Links
Gene Details
- Gene
- SFTA3
- Gene Name
- surfactant associated 3
View Full Product Details