Product Details
- SNP ID
-
rs201043457
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:67823597 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTATCACAAGAGCTGTGTGACCG[A/T]CTGAGTAGACATCAGATCCTTACCT
- Phenotype
-
MIM: 602948
MIM: 612012
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RAD51B
PubMed Links
Gene Details
- Gene
- RAD51B
- Gene Name
- RAD51 paralog B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321809.1 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_001308738.1 |
NM_001321810.1 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_001308739.1 |
NM_001321812.1 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_001308741.1 |
NM_001321814.1 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_001308743.1 |
NM_001321815.1 |
87 |
UTR 5 |
|
|
NP_001308744.1 |
NM_001321817.1 |
87 |
UTR 5 |
|
|
NP_001308746.1 |
NM_001321818.1 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_001308747.1 |
NM_001321819.1 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_001308748.1 |
NM_001321821.1 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_001308750.1 |
NM_002877.5 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_002868.1 |
NM_133509.3 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_598193.2 |
NM_133510.3 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
NP_598194.1 |
XM_011537050.2 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
XP_011535352.1 |
XM_011537051.2 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
XP_011535353.1 |
XM_017021545.1 |
87 |
Silent Mutation |
CGA,CGT |
R18R |
XP_016877034.1 |
XM_017021546.1 |
87 |
Intron |
|
|
XP_016877035.1 |
XM_017021547.1 |
87 |
Intron |
|
|
XP_016877036.1 |
XM_017021548.1 |
87 |
Intron |
|
|
XP_016877037.1 |
- Gene
- ZFYVE26
- Gene Name
- zinc finger FYVE-type containing 26
There are no transcripts associated with this gene.
View Full Product Details