Product Details

SNP ID
rs200936790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20117822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATATTTCTCCTTCACTTACTGG[G/T]TGGGGTTGAAATGGTACTGTTGGTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4K17 PubMed Links

Gene Details

Gene
OR4K17
Gene Name
olfactory receptor family 4 subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004715.1 416 Missense Mutation GGT,GTT G139V NP_001004715.1

View Full Product Details