Product Details

SNP ID
rs200937141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34753647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCTTTTGCTTCACTTGTGTTA[C/T]GAGGGTTGTATTCAAAGCAGTTCGA
Phenotype
MIM: 605680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAZ1A PubMed Links

Gene Details

Gene
BAZ1A
Gene Name
bromodomain adjacent to zinc finger domain 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013448.2 3227 Missense Mutation CAT,CGT H1511R NP_038476.2
NM_182648.1 3227 Missense Mutation CAT,CGT H1479R NP_872589.1
XM_011536374.1 3227 Missense Mutation CAT,CGT H1418R XP_011534676.1
XM_011536376.2 3227 Missense Mutation CAT,CGT H1034R XP_011534678.2
XM_017020942.1 3227 Missense Mutation CAT,CGT H1002R XP_016876431.1

View Full Product Details