Product Details

SNP ID
rs201622896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75003667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCCCAACTCCAGTCCAACATCA[C/T]TGAGGCGATTAATGAGCTGCTAGTG
Phenotype
MIM: 606454
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2B2 PubMed Links

Gene Details

Gene
EIF2B2
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014239.3 483 Missense Mutation ACT,ATT T134I NP_055054.1

View Full Product Details