Product Details

SNP ID
rs200472203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23049722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAACTTCATCTCTGTCCAGGGCC[C/T]GGATGACCTGAATCAGCTGGGAGGA
Phenotype
MIM: 604562 MIM: 611137
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACIN1 PubMed Links

Gene Details

Gene
ACIN1
Gene Name
apoptotic chromatin condensation inducer 1
There are no transcripts associated with this gene.

Gene
CDH24
Gene Name
cadherin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022478.3 1876 Missense Mutation CAG,CGG Q539R NP_071923.2
NM_144985.3 1876 Missense Mutation CAG,CGG Q501R NP_659422.2
XM_011537089.1 1876 UTR 3 XP_011535391.1
Gene
LOC105370705
Gene Name
collagen alpha-2(I) chain-like
There are no transcripts associated with this gene.

Gene
PSMB11
Gene Name
proteasome subunit beta 11
There are no transcripts associated with this gene.

View Full Product Details