Product Details

SNP ID
rs200759183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100538344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGAAGCTGTCGGCCTTGTAGCT[A/G]GCGTAGAGCGGGCTGGCGCGGCCGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BEGAIN PubMed Links

Gene Details

Gene
BEGAIN
Gene Name
brain enriched guanylate kinase associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159531.1 1695 Silent Mutation GCC,GCT A469A NP_001153003.1
NM_020836.3 1695 Silent Mutation GCC,GCT A469A NP_065887.1
XM_005267919.3 1695 Silent Mutation GCC,GCT A482A XP_005267976.1
XM_005267920.4 1695 Silent Mutation GCC,GCT A481A XP_005267977.1
XM_005267921.3 1695 Silent Mutation GCC,GCT A469A XP_005267978.1
XM_011537031.1 1695 Silent Mutation GCC,GCT A488A XP_011535333.1
XM_017021521.1 1695 Silent Mutation GCC,GCT A469A XP_016877010.1
XM_017021522.1 1695 Silent Mutation GCC,GCT A460A XP_016877011.1
Gene
WDR25
Gene Name
WD repeat domain 25
There are no transcripts associated with this gene.

View Full Product Details