Product Details

SNP ID
rs201121939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24249535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGTCACTGTTTCATTGCCTCC[A/G]ATGTCCCTGTGGGCAGAGACAAGGT
Phenotype
MIM: 190195 MIM: 604319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TGM1 PubMed Links

Gene Details

Gene
TGM1
Gene Name
transglutaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000359.2 2356 Silent Mutation ATC,ATT I744I NP_000350.1
Gene
TINF2
Gene Name
TERF1 interacting nuclear factor 2
There are no transcripts associated with this gene.

View Full Product Details