Product Details

SNP ID
rs201880365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20117690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACTCCCATGTATTTTCTCCTTG[A/G]TAATCTCTCTTTTGTAGATATGACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4K17 PubMed Links

Gene Details

Gene
OR4K17
Gene Name
olfactory receptor family 4 subfamily K member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004715.1 284 Missense Mutation GAT,GGT D95G NP_001004715.1

View Full Product Details