Product Details
- SNP ID
-
rs200409299
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:31106685 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTAAGTGGAGGCCCAGAACCTGAT[G/T]GATTCTTCAGCACTAGTTCCTGTAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HECTD1
PubMed Links
Gene Details
- Gene
- HECTD1
- Gene Name
- HECT domain E3 ubiquitin protein ligase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015382.3 |
7067 |
Missense Mutation |
CAA,CCA |
Q2396P |
NP_056197.3 |
XM_005267502.2 |
7067 |
Missense Mutation |
CAA,CCA |
Q2396P |
XP_005267559.2 |
XM_011536619.2 |
7067 |
Missense Mutation |
CAA,CCA |
Q2394P |
XP_011534921.1 |
XM_011536621.2 |
7067 |
Missense Mutation |
CAA,CCA |
Q2117P |
XP_011534923.1 |
XM_011536622.1 |
7067 |
Missense Mutation |
CAA,CCA |
Q1846P |
XP_011534924.1 |
XM_017021148.1 |
7067 |
Missense Mutation |
CAA,CCA |
Q2349P |
XP_016876637.1 |
XM_017021149.1 |
7067 |
Missense Mutation |
CAA,CCA |
Q2347P |
XP_016876638.1 |
XM_017021150.1 |
7067 |
Missense Mutation |
CAA,CCA |
Q2335P |
XP_016876639.1 |
XM_017021151.1 |
7067 |
Missense Mutation |
CAA,CCA |
Q2117P |
XP_016876640.1 |
XM_017021152.1 |
7067 |
Missense Mutation |
CAA,CCA |
Q2115P |
XP_016876641.1 |
XM_017021153.1 |
7067 |
Missense Mutation |
CAA,CCA |
Q2115P |
XP_016876642.1 |
View Full Product Details