Product Details

SNP ID
rs201349357
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52432392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATTACTTCTATTCCAATGCTTT[A/C]TTCTCAGAGTTTCAATCGGTTCTTT
Phenotype
MIM: 616179
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
TXNDC16 PubMed Links

Gene Details

Gene
TXNDC16
Gene Name
thioredoxin domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160047.1 2824 Missense Mutation AGA,ATA R792I NP_001153519.1
NM_020784.2 2824 Missense Mutation AGA,ATA R797I NP_065835.2
XM_017021505.1 2824 Intron XP_016876994.1

View Full Product Details