Product Details

SNP ID
rs199839288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90263357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCCGGTTAAAATTACTGAAGT[C/T]AGAGAGAATTAAAGACTATCTTCTC
Phenotype
MIM: 602706
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMC1 PubMed Links

Gene Details

Gene
PSMC1
Gene Name
proteasome 26S subunit, ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002802.2 1761 Intron NP_002793.2
XM_017021470.1 1761 UTR 5 XP_016876959.1
XM_017021471.1 1761 Intron XP_016876960.1

View Full Product Details