Product Details

SNP ID
rs201896588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22923114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGTACAGCTTGGAGGAAGAGAT[A/G]GGAGCCAGAAAGGAAGTGTACTCCC
Phenotype
MIM: 604045
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101926933 PubMed Links

Gene Details

Gene
LOC101926933
Gene Name
uncharacterized LOC101926933
There are no transcripts associated with this gene.

Gene
PRMT5
Gene Name
protein arginine methyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039619.2 1269 Silent Mutation CCC,CCT P457P NP_001034708.1
NM_001282953.1 1269 Silent Mutation CCC,CCT P413P NP_001269882.1
NM_001282954.1 1269 Silent Mutation CCC,CCT P368P NP_001269883.1
NM_001282955.1 1269 Silent Mutation CCC,CCT P430P NP_001269884.1
NM_001282956.1 1269 Silent Mutation CCC,CCT P303P NP_001269885.1
NM_006109.4 1269 Silent Mutation CCC,CCT P474P NP_006100.2
XM_017020927.1 1269 Silent Mutation CCC,CCT P368P XP_016876416.1
Gene
PRMT5-AS1
Gene Name
PRMT5 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RBM23
Gene Name
RNA binding motif protein 23
There are no transcripts associated with this gene.

View Full Product Details