Product Details

SNP ID
rs200925018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23348258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGACTGAGCCTCCTCAATCTG[C/T]CGCTTCACTATCAGCCACCGTGCGG
Phenotype
MIM: 609906 MIM: 611461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EFS PubMed Links

Gene Details

Gene
EFS
Gene Name
embryonal Fyn-associated substrate
There are no transcripts associated with this gene.

Gene
SLC22A17
Gene Name
solute carrier family 22 member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289050.1 1245 Silent Mutation NP_001275979.1
NM_016609.4 1245 Silent Mutation NP_057693.3
NM_020372.3 1245 Silent Mutation NP_065105.2
XM_005267747.4 1245 Silent Mutation XP_005267804.1
XM_005267748.4 1245 Silent Mutation XP_005267805.1
XM_017021361.1 1245 Silent Mutation XP_016876850.1
XM_017021362.1 1245 Silent Mutation XP_016876851.1

View Full Product Details