Product Details

SNP ID
rs202122792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102923729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCAGCACTGACCCAGGCGGTCT[C/T]CAAACTCTGGGTCCCCAACACGGAC
Phenotype
MIM: 605799 MIM: 614062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMN PubMed Links

Gene Details

Gene
AMN
Gene Name
amnion associated transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030943.3 95 Missense Mutation TCC,TTC S21F NP_112205.2
XM_011537202.2 95 UTR 5 XP_011535504.1
XM_011537203.2 95 UTR 5 XP_011535505.1
Gene
CDC42BPB
Gene Name
CDC42 binding protein kinase beta
There are no transcripts associated with this gene.

View Full Product Details