Product Details
- SNP ID
-
rs199582609
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:91234555 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAAGCACACGCGGTGCTGGTTCT[C/T]GTCCTCGATGACCTCCTCGTGCATC
- Phenotype
-
MIM: 601404
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C14orf159
PubMed Links
Gene Details
- Gene
- C14orf159
- Gene Name
- chromosome 14 open reading frame 159
There are no transcripts associated with this gene.
- Gene
- GPR68
- Gene Name
- G protein-coupled receptor 68
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001177676.1 |
792 |
Missense Mutation |
AAG,GAG |
K166E |
NP_001171147.1 |
NM_003485.3 |
792 |
Missense Mutation |
AAG,GAG |
K166E |
NP_003476.3 |
XM_005268110.4 |
792 |
Missense Mutation |
AAG,GAG |
K176E |
XP_005268167.1 |
XM_005268111.3 |
792 |
Missense Mutation |
AAG,GAG |
K176E |
XP_005268168.1 |
XM_005268112.3 |
792 |
Missense Mutation |
AAG,GAG |
K176E |
XP_005268169.1 |
XM_006720262.3 |
792 |
Missense Mutation |
AAG,GAG |
K176E |
XP_006720325.1 |
XM_011537196.2 |
792 |
Missense Mutation |
AAG,GAG |
K176E |
XP_011535498.1 |
XM_011537197.2 |
792 |
Missense Mutation |
AAG,GAG |
K176E |
XP_011535499.1 |
XM_011537198.2 |
792 |
Missense Mutation |
AAG,GAG |
K176E |
XP_011535500.1 |
XM_011537199.2 |
792 |
Missense Mutation |
AAG,GAG |
K176E |
XP_011535501.1 |
View Full Product Details