Product Details

SNP ID
rs201395244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67521454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCAGTTCGGCCTCTCGCACTTCA[A/G]CTGGCCGGTGCAGGGCGCCAACTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C15orf61 PubMed Links

Gene Details

Gene
C15orf61
Gene Name
chromosome 15 open reading frame 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143936.1 271 Missense Mutation AAC,AGC N69S NP_001137408.1
Gene
IQCH-AS1
Gene Name
IQCH antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details