Product Details

SNP ID
rs201775551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65076925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCTTCTTCCGAGGCCTCTTCC[A/G]CTCCGGCATGCGGGAGACCCGCGCA
Phenotype
MIM: 613727
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KBTBD13 PubMed Links

Gene Details

Gene
KBTBD13
Gene Name
kelch repeat and BTB domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101362.2 110 Missense Mutation CAC,CGC H37R NP_001094832.1
Gene
RASL12
Gene Name
RAS like family 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307930.1 110 Intron NP_001294859.1
NM_016563.3 110 Intron NP_057647.1
XM_005254434.4 110 Intron XP_005254491.1
XM_011521660.2 110 Intron XP_011519962.1
XM_017022296.1 110 Intron XP_016877785.1

View Full Product Details