Product Details

SNP ID
rs201330867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68208164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTGCCGACTGCTGACGTGAAG[A/G]GTGTAGAAAGCCCAGGGCTCAGGGA
Phenotype
MIM: 606725
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALML4 PubMed Links

Gene Details

Gene
CALML4
Gene Name
calmodulin like 4
There are no transcripts associated with this gene.

Gene
CLN6
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017882.2 1070 Silent Mutation ACC,ACT T304T NP_060352.1

View Full Product Details