Product Details

SNP ID
rs199871287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73873754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACAGCCTCTCTGCCAGACAGTC[A/T]GCCTCCTTCATCCTGGTGCGTGTTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TBC1D21 PubMed Links

Gene Details

Gene
TBC1D21
Gene Name
TBC1 domain family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286434.1 198 Silent Mutation TCA,TCT S15S NP_001273363.1
NM_153356.2 198 Silent Mutation TCA,TCT S15S NP_699187.1
XM_006720409.3 198 Silent Mutation TCA,TCT S15S XP_006720472.1
XM_006720410.3 198 Silent Mutation TCA,TCT S15S XP_006720473.1
XM_006720411.3 198 UTR 5 XP_006720474.1
XM_011521281.2 198 Silent Mutation TCA,TCT S15S XP_011519583.1
XM_011521282.2 198 Silent Mutation TCA,TCT S15S XP_011519584.1
XM_011521283.2 198 Silent Mutation TCA,TCT S15S XP_011519585.1
XM_011521284.2 198 Intron XP_011519586.1

View Full Product Details