Product Details

SNP ID
rs200031104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67824946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGAAAGCTCAAGGAGTGGGCG[A/C]CTGGAAGCTAAGAAGTGGATCCCAA
Phenotype
MIM: 611273
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929076 PubMed Links

Gene Details

Gene
LOC101929076
Gene Name
uncharacterized LOC101929076
There are no transcripts associated with this gene.

Gene
SKOR1
Gene Name
SKI family transcriptional corepressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258024.1 582 Silent Mutation CGA,CGC R194R NP_001244953.1
XM_017022173.1 582 Intron XP_016877662.1

View Full Product Details