Product Details
- SNP ID
-
rs200658675
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:63277633 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTTCCGCGGTGGCCATGACTGCG[A/G]CCGTGTTCTTCGGCTGCGCCTTCAT
- Phenotype
-
MIM: 607630
MIM: 613532
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APH1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs189409411] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APH1B
- Gene Name
- aph-1 homolog B, gamma-secretase subunit
- Gene
- RAB8B
- Gene Name
- RAB8B, member RAS oncogene family
There are no transcripts associated with this gene.
View Full Product Details