Product Details

SNP ID
rs200440894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51723647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTTGCATAGGGACTTTCTTCA[A/T]CTCTGAAAGAAAATAAATACAGCAT
Phenotype
MIM: 611796
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LYSMD2 PubMed Links

Gene Details

Gene
LYSMD2
Gene Name
LysM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143917.1 501 Missense Mutation GAT,GTT D112V NP_001137389.1
NM_153374.2 501 Missense Mutation GAT,GTT D203V NP_699205.1
XM_017022057.1 501 Missense Mutation GAT,GTT D112V XP_016877546.1
Gene
SCG3
Gene Name
secretogranin III
There are no transcripts associated with this gene.

View Full Product Details