Product Details

SNP ID
rs200597403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84680827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGTCAAAAACTTGATATGCCCA[C/T]TTTGCCTTAAAAGAGTAAAGGAAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC11A PubMed Links

Gene Details

Gene
SEC11A
Gene Name
SEC11 homolog A, signal peptidase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271918.1 838 Missense Mutation AAT,AGT N80S NP_001258847.1
NM_001271919.1 838 Intron NP_001258848.1
NM_001271920.1 838 Missense Mutation AAT,AGT N106S NP_001258849.1
NM_001271921.1 838 Missense Mutation AAT,AGT N56S NP_001258850.1
NM_001271922.1 838 Missense Mutation AAT,AGT N106S NP_001258851.1
NM_014300.3 838 Missense Mutation AAT,AGT N106S NP_055115.1

View Full Product Details