Product Details
- SNP ID
-
rs200785525
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:45094679 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGTTCAGCACTTTCTGGAAGTGCC[A/G]CTCGCAGATGTACTGGGGGCACAGG
- Phenotype
-
MIM: 606759
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DUOX2
PubMed Links
Gene Details
- Gene
- DUOX2
- Gene Name
- dual oxidase 2
View Full Product Details