Product Details

SNP ID
rs200129131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:647442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGGCCAAGGCTTGTGTTCAAT[C/T]GTGTGAATGGCCGGCGGGCCCCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100287175 PubMed Links

Gene Details

Gene
LOC100287175
Gene Name
uncharacterized LOC100287175
There are no transcripts associated with this gene.

Gene
LOC105371038
Gene Name
uncharacterized LOC105371038
There are no transcripts associated with this gene.

Gene
MCRIP2
Gene Name
MAPK regulated corepressor interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138418.2 Intron NP_612427.2
Gene
WDR90
Gene Name
WD repeat domain 90
There are no transcripts associated with this gene.

View Full Product Details