Product Details
- SNP ID
-
rs200852524
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:56639270 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTTCTCTTCCTTGCAGGTGGCT[C/G]CTGCACCTGCACTGGCTCCTGCAAA
- Phenotype
-
MIM: 156350
MIM: 156356
MIM: 156357
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MT1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11647171] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MT1A
- Gene Name
- metallothionein 1A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005946.2 |
108 |
Missense Mutation |
TCC,TGC |
S12C |
NP_005937.2 |
- Gene
- MT1DP
- Gene Name
- metallothionein 1D, pseudogene
There are no transcripts associated with this gene.
- Gene
- MT1JP
- Gene Name
- metallothionein 1J, pseudogene
There are no transcripts associated with this gene.
- Gene
- MT1M
- Gene Name
- metallothionein 1M
There are no transcripts associated with this gene.
View Full Product Details