Product Details

SNP ID
rs199544373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:854561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCCCGGGCAGAGGCCACCCAC[A/G]GTCCCTGAAGTAGGGCCTCAGCTCC
Phenotype
MIM: 611761
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMF1 PubMed Links

Gene Details

Gene
LMF1
Gene Name
lipase maturation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022773.2 2258 Missense Mutation CGT,TGT R559C NP_073610.2
XM_006720928.2 2258 Missense Mutation CGT,TGT R450C XP_006720991.1
XM_006720929.3 2258 Missense Mutation CGT,TGT R426C XP_006720992.1
XM_011522614.2 2258 Intron XP_011520916.1
XM_011522615.1 2258 Intron XP_011520917.1
XM_011522617.2 2258 Missense Mutation CGT,TGT R342C XP_011520919.1
XM_011522618.2 2258 Intron XP_011520920.1
XM_017023575.1 2258 Intron XP_016879064.1
XM_017023576.1 2258 Missense Mutation CGT,TGT R481C XP_016879065.1
XM_017023577.1 2258 Missense Mutation CGT,TGT R426C XP_016879066.1
XM_017023578.1 2258 Missense Mutation CGT,TGT R347C XP_016879067.1
XM_017023579.1 2258 Missense Mutation CGT,TGT R342C XP_016879068.1
XM_017023580.1 2258 Missense Mutation CGT,TGT R342C XP_016879069.1
XM_017023581.1 2258 Intron XP_016879070.1

View Full Product Details