Product Details

SNP ID
rs199517696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67829965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTTTTCTCTGCCTCTTCAACT[A/C]CATCGTGTAAGGGCTCTACTTCATC
Phenotype
MIM: 611510 MIM: 607720
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CENPT PubMed Links

Gene Details

Gene
CENPT
Gene Name
centromere protein T
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025082.3 1390 Missense Mutation NP_079358.3
XM_006721277.2 1390 Missense Mutation XP_006721340.1
XM_011523344.1 1390 Missense Mutation XP_011521646.1
XM_011523345.1 1390 Missense Mutation XP_011521647.1
XM_017023712.1 1390 Missense Mutation XP_016879201.1
XM_017023713.1 1390 Missense Mutation XP_016879202.1
XM_017023714.1 1390 Missense Mutation XP_016879203.1
XM_017023715.1 1390 Missense Mutation XP_016879204.1
XM_017023716.1 1390 Missense Mutation XP_016879205.1
XM_017023717.1 1390 Missense Mutation XP_016879206.1
Gene
TSNAXIP1
Gene Name
translin associated factor X interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details