Product Details

SNP ID
rs200039844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29901864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGCTTTTCCGTTCTCCTGCCTG[C/G]GGCCCGGCTCGAGGGCCGCTGTGGG
Phenotype
MIM: 608947 MIM: 616667
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASPHD1 PubMed Links

Gene Details

Gene
ASPHD1
Gene Name
aspartate beta-hydroxylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181718.3 1039 Missense Mutation GCG,GGG A298G NP_859069.2
XM_017023107.1 1039 Missense Mutation GCG,GGG A147G XP_016878596.1
Gene
KCTD13
Gene Name
potassium channel tetramerization domain containing 13
There are no transcripts associated with this gene.

Gene
SEZ6L2
Gene Name
seizure related 6 homolog like 2
There are no transcripts associated with this gene.

View Full Product Details