Product Details

SNP ID
rs201150509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1691868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTAAAGGAAGTGGTATCTTTGA[C/T]GAATCAACCCCCGTGCAGACTCGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HN1L PubMed Links

Gene Details

Gene
HN1L
Gene Name
hematological and neurological expressed 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144570.2 255 Silent Mutation GAC,GAT D73D NP_653171.1

View Full Product Details