Product Details

SNP ID
rs200962346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2151583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGTTTCAGAACAAAGTTCTGGA[A/C]GTGGATGGTGTGAAGGTGAAGCTGC
Phenotype
MIM: 605455 MIM: 606692
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RAB26 PubMed Links

Gene Details

Gene
RAB26
Gene Name
RAB26, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308053.1 494 Silent Mutation GAA,GAC E41D NP_001294982.1
NM_014353.4 494 Silent Mutation GAA,GAC E107D NP_055168.2
XM_011522448.1 494 Missense Mutation GAA,GAC E107D XP_011520750.1
XM_011522450.2 494 UTR 5 XP_011520752.1
Gene
SNHG19
Gene Name
small nucleolar RNA host gene 19
There are no transcripts associated with this gene.

Gene
SNORD60
Gene Name
small nucleolar RNA, C/D box 60
There are no transcripts associated with this gene.

Gene
TRAF7
Gene Name
TNF receptor associated factor 7
There are no transcripts associated with this gene.

View Full Product Details