Product Details
- SNP ID
-
rs200051096
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:727581 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTCATCGAGGAGCTCACGCGCGA[C/G]GCGGTGGCCGTGGACGTGGCTGTGC
- Phenotype
-
MIM: 614666
MIM: 610998
MIM: 611118
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC78
PubMed Links
Gene Details
- Gene
- CCDC78
- Gene Name
- coiled-coil domain containing 78
- Gene
- FAM173A
- Gene Name
- family with sequence similarity 173 member A
There are no transcripts associated with this gene.
- Gene
- HAGHL
- Gene Name
- hydroxyacylglutathione hydrolase-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290137.1 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
NP_001277066.1 |
NM_001290139.1 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
NP_001277068.1 |
NM_001323635.1 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
NP_001310564.1 |
NM_001323636.1 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
NP_001310565.1 |
NM_032304.3 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
NP_115680.1 |
XM_005255631.4 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
XP_005255688.1 |
XM_005255632.1 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
XP_005255689.1 |
XM_011522711.1 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
XP_011521013.1 |
XM_011522712.1 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
XP_011521014.1 |
XM_017023773.1 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
XP_016879262.1 |
XM_017023774.1 |
570 |
Missense Mutation |
GAC,GAG |
D24E |
XP_016879263.1 |
- Gene
- METRN
- Gene Name
- meteorin, glial cell differentiation regulator
There are no transcripts associated with this gene.
- Gene
- NARFL
- Gene Name
- nuclear prelamin A recognition factor like
There are no transcripts associated with this gene.
View Full Product Details