Product Details
- SNP ID
-
rs200446825
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28826896 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGATTCAGCCATTGCCATGAACTC[A/G]AAAGTGAATGGGGAACACAAAGAGA
- Phenotype
-
MIM: 607931
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN2L
PubMed Links
Gene Details
- Gene
- ATXN2L
- Gene Name
- ataxin 2 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308230.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
NP_001295159.1 |
NM_007245.3 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
NP_009176.2 |
NM_017492.3 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
NP_059867.3 |
NM_145714.2 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
NP_663760.1 |
NM_148414.2 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
NP_680780.1 |
NM_148415.2 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
NP_680781.1 |
NM_148416.2 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
NP_680782.1 |
XM_005255061.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255118.1 |
XM_005255062.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255119.1 |
XM_005255063.3 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255120.1 |
XM_005255064.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255121.1 |
XM_005255065.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255122.1 |
XM_005255066.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255123.1 |
XM_005255067.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255124.1 |
XM_005255068.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255125.1 |
XM_005255069.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255126.1 |
XM_005255070.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255127.1 |
XM_005255071.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255128.1 |
XM_005255074.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255131.1 |
XM_005255075.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255132.1 |
XM_005255076.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_005255133.1 |
XM_005255077.1 |
863 |
Silent Mutation |
TCA,TCG |
S157S |
XP_005255134.1 |
XM_006721007.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_006721070.1 |
XM_006721008.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_006721071.1 |
XM_006721009.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_006721072.1 |
XM_006721010.2 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_006721073.1 |
XM_006721011.1 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_006721074.1 |
XM_006721012.3 |
863 |
Intron |
|
|
XP_006721075.1 |
XM_011545719.2 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_011544021.1 |
XM_011545721.2 |
863 |
Silent Mutation |
TCA,TCG |
S217S |
XP_011544023.1 |
XM_011545722.1 |
863 |
Intron |
|
|
XP_011544024.1 |
XM_017022891.1 |
863 |
Silent Mutation |
TCA,TCG |
S157S |
XP_016878380.1 |
XM_017022892.1 |
863 |
Intron |
|
|
XP_016878381.1 |
View Full Product Details