Product Details

SNP ID
rs200544857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89697383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGAGCAGAGTGTCCATCTGTGCA[C/T]GCTGTAGCCACAAGCGCCGCTGGGC
Phenotype
MIM: 603464
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK10 PubMed Links

Gene Details

Gene
CDK10
Gene Name
cyclin dependent kinase 10
There are no transcripts associated with this gene.

Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152339.3 1730 Missense Mutation CAT,CGT H409R NP_689552.2
XM_005256279.4 1730 Missense Mutation CAT,CGT H409R XP_005256336.1
Gene
VPS9D1
Gene Name
VPS9 domain containing 1
There are no transcripts associated with this gene.

View Full Product Details