Product Details
- SNP ID
-
rs199564998
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:67003084 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTCTTGCAGATGATGTCACCCCT[A/G]GCCTCGGGTCTCTTCCATCGGGCCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CES4A
PubMed Links
Gene Details
- Gene
- CES4A
- Gene Name
- carboxylesterase 4A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001190201.1 |
1018 |
Silent Mutation |
CTA,CTG |
L137L |
NP_001177130.1 |
NM_001190202.1 |
1018 |
Silent Mutation |
CTA,CTG |
L141L |
NP_001177131.1 |
NM_001318506.1 |
1018 |
Silent Mutation |
CTA,CTG |
L41L |
NP_001305435.1 |
NM_173815.6 |
1018 |
Silent Mutation |
CTA,CTG |
L235L |
NP_776176.5 |
XM_005255896.4 |
1018 |
Silent Mutation |
CTA,CTG |
L258L |
XP_005255953.1 |
XM_011523021.2 |
1018 |
Silent Mutation |
CTA,CTG |
L302L |
XP_011521323.1 |
XM_011523023.2 |
1018 |
Silent Mutation |
CTA,CTG |
L298L |
XP_011521325.1 |
XM_011523024.2 |
1018 |
Silent Mutation |
CTA,CTG |
L302L |
XP_011521326.1 |
XM_011523025.2 |
1018 |
Silent Mutation |
CTA,CTG |
L302L |
XP_011521327.1 |
XM_011523026.2 |
1018 |
Silent Mutation |
CTA,CTG |
L262L |
XP_011521328.1 |
XM_011523027.2 |
1018 |
Silent Mutation |
CTA,CTG |
L302L |
XP_011521329.1 |
XM_011523028.2 |
1018 |
Silent Mutation |
CTA,CTG |
L250L |
XP_011521330.1 |
XM_011523029.2 |
1018 |
Silent Mutation |
CTA,CTG |
L302L |
XP_011521331.1 |
XM_011523030.2 |
1018 |
Silent Mutation |
CTA,CTG |
L258L |
XP_011521332.1 |
XM_017023160.1 |
1018 |
Silent Mutation |
CTA,CTG |
L298L |
XP_016878649.1 |
XM_017023161.1 |
1018 |
Silent Mutation |
CTA,CTG |
L298L |
XP_016878650.1 |
XM_017023162.1 |
1018 |
Silent Mutation |
CTA,CTG |
L302L |
XP_016878651.1 |
XM_017023163.1 |
1018 |
Silent Mutation |
CTA,CTG |
L302L |
XP_016878652.1 |
XM_017023164.1 |
1018 |
Silent Mutation |
CTA,CTG |
L298L |
XP_016878653.1 |
XM_017023165.1 |
1018 |
Silent Mutation |
CTA,CTG |
L298L |
XP_016878654.1 |
XM_017023166.1 |
1018 |
Silent Mutation |
CTA,CTG |
L302L |
XP_016878655.1 |
XM_017023167.1 |
1018 |
Silent Mutation |
CTA,CTG |
L137L |
XP_016878656.1 |
XM_017023168.1 |
1018 |
Silent Mutation |
CTA,CTG |
L137L |
XP_016878657.1 |
View Full Product Details