Product Details

SNP ID
rs200058857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67646761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTGTGTGACTACAATGGCTTC[C/G]CTTTCCGAGAGGAGATTCAGTGGGT
Phenotype
MIM: 610859 MIM: 604167
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CARMIL2 PubMed Links

Gene Details

Gene
CARMIL2
Gene Name
capping protein regulator and myosin 1 linker 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013838.1 757 Missense Mutation CCT,GCT P172A NP_001013860.1
NM_001317026.1 757 Missense Mutation CCT,GCT P172A NP_001303955.1
XM_011522874.1 757 Missense Mutation CCT,GCT P172A XP_011521176.1
XM_011522875.2 757 Missense Mutation CCT,GCT P172A XP_011521177.2
XM_017022953.1 757 Missense Mutation CCT,GCT P172A XP_016878442.1
Gene
CTCF
Gene Name
CCCTC-binding factor
There are no transcripts associated with this gene.

View Full Product Details