Product Details

SNP ID
rs200157828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29807331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGTCGCGTCTGCCTTGGAGAA[A/G]AAGACAAAGAGCAAGGGGCCCTACA
Phenotype
MIM: 603213 MIM: 600999 MIM: 612033 MIM: 614386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
There are no transcripts associated with this gene.

Gene
MAZ
Gene Name
MYC associated zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042539.2 526 Silent Mutation AAA,AAG K182K NP_001036004.1
NM_001276275.1 526 Silent Mutation AAA,AAG K159K NP_001263204.1
NM_001276276.1 526 Intron NP_001263205.1
NM_002383.3 526 Silent Mutation AAA,AAG K182K NP_002374.2
XM_006721047.3 526 Silent Mutation AAA,AAG K159K XP_006721110.1
XM_006721048.3 526 Silent Mutation AAA,AAG K182K XP_006721111.1
Gene
PAGR1
Gene Name
PAXIP1 associated glutamate rich protein 1
There are no transcripts associated with this gene.

Gene
PRRT2
Gene Name
proline rich transmembrane protein 2
There are no transcripts associated with this gene.

View Full Product Details