Product Details
- SNP ID
-
rs201628798
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2771523 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGGGTCCGTCTTGGGGTTCCCT[C/T]GTTGAACATGCTGTCAAACCAGGAC
- Phenotype
-
MIM: 606032
MIM: 600787
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SRRM2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4036] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SRRM2
- Gene Name
- serine/arginine repetitive matrix 2
- Gene
- TCEB2
- Gene Name
- transcription elongation factor B subunit 2
View Full Product Details