Product Details
- SNP ID
-
rs202215040
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:20933405 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGATTTTGGGGAGAGATTCCCCAA[A/T]CTGCATCGTTTTCCTGTCCCAACGG
- Phenotype
-
MIM: 603334
MIM: 614709
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DNAH3
PubMed Links
Gene Details
- Gene
- DNAH3
- Gene Name
- dynein axonemal heavy chain 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017539.2 |
12678 |
Missense Mutation |
ATT,TTT |
I4034F |
NP_060009.1 |
XM_006721062.1 |
12678 |
Missense Mutation |
ATT,TTT |
I3988F |
XP_006721125.1 |
XM_011545882.2 |
12678 |
Missense Mutation |
ATT,TTT |
I3995F |
XP_011544184.1 |
XM_011545883.1 |
12678 |
Missense Mutation |
ATT,TTT |
I3995F |
XP_011544185.1 |
XM_011545885.2 |
12678 |
Intron |
|
|
XP_011544187.1 |
XM_011545886.2 |
12678 |
Missense Mutation |
ATT,TTT |
I3658F |
XP_011544188.1 |
XM_011545888.2 |
12678 |
Missense Mutation |
ATT,TTT |
I3129F |
XP_011544190.1 |
XM_011545889.2 |
12678 |
Missense Mutation |
ATT,TTT |
I3129F |
XP_011544191.1 |
XM_017023426.1 |
12678 |
Missense Mutation |
ATT,TTT |
I3950F |
XP_016878915.1 |
XM_017023427.1 |
12678 |
Missense Mutation |
ATT,TTT |
I3914F |
XP_016878916.1 |
XM_017023428.1 |
12678 |
Missense Mutation |
ATT,TTT |
I3802F |
XP_016878917.1 |
XM_017023429.1 |
12678 |
Intron |
|
|
XP_016878918.1 |
XM_017023430.1 |
12678 |
Missense Mutation |
ATT,TTT |
I2013F |
XP_016878919.1 |
XM_017023431.1 |
12678 |
Intron |
|
|
XP_016878920.1 |
XM_017023432.1 |
12678 |
Missense Mutation |
ATT,TTT |
I4048F |
XP_016878921.1 |
- Gene
- LYRM1
- Gene Name
- LYR motif containing 1
There are no transcripts associated with this gene.
View Full Product Details