Product Details

SNP ID
rs201932925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2818868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGTCTGCACCTGTCACCTACA[C/T]TAAACACATCCAGCCCATCTGTCTC
Phenotype
MIM: 608159
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS21 PubMed Links

Gene Details

Gene
PRSS21
Gene Name
protease, serine 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270452.1 475 Missense Mutation ACT,ATT T148I NP_001257381.1
NM_006799.3 475 Missense Mutation ACT,ATT T150I NP_006790.1
NM_144956.2 475 Missense Mutation ACT,ATT T148I NP_659205.1
NM_144957.2 475 Missense Mutation ACT,ATT T150I NP_659206.1
XM_017022875.1 475 Missense Mutation ACT,ATT T148I XP_016878364.1
XM_017022876.1 475 Missense Mutation ACT,ATT T150I XP_016878365.1
XM_017022877.1 475 Missense Mutation ACT,ATT T148I XP_016878366.1
XM_017022878.1 475 Missense Mutation ACT,ATT T150I XP_016878367.1

View Full Product Details