Product Details

SNP ID
rs273585633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88428490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACCCCTGCCCTGCTACCAGGG[C/T]CAGCCAGGTGGCCTGAACCGCCACA
Phenotype
MIM: 612078
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF469 PubMed Links

Gene Details

Gene
ZNF469
Gene Name
zinc finger protein 469
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127464.2 1213 Silent Mutation GGC,GGT G340G NP_001120936.2
XM_017023784.1 1213 Silent Mutation GGC,GGT G340G XP_016879273.1
XM_017023785.1 1213 Silent Mutation GGC,GGT G340G XP_016879274.1

View Full Product Details